Assay Details
Gene Symbol: Gnb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_030987.2 | NP_112249.2 | 7 - 8 | 573 | 72 | |
XM_006239517.2 | XP_006239579.1 | 7 - 8 | 725 | 72 | ||
XM_006239518.2 | XP_006239580.1 | 7 - 8 | 725 | 72 | ||
XM_006239519.2 | XP_006239581.1 | 7 - 8 | 725 | 72 | ||
XM_006239520.2 | XP_006239582.1 | 7 - 8 | 725 | 72 | ||
XM_008764335.1 | XP_008762557.1 | 6 - 7 | 608 | 72 | ||
GenBank mRNA | AY552805.1 | - | 5 - 6 | 425 | 72 | |
BC078809.1 | - | 7 - 8 | 573 | 72 | ||
DQ120483.1 | - | 2 - 3 | 178 | 72 | ||
DQ120484.1 | - | 2 - 3 | 178 | 72 | ||
U34958.1 | - | 5 - 6 | 425 | 72 | ||
U88324.1 | - | 5 - 6 | 462 | 72 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
