Assay Details
Gene Symbol: Rpl5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031099.1 | NP_112361.1 | 5 - 6 | 826 | 106 | |
GenBank mRNA | BC060561.1 | - | 5 - 6 | 773 | 106 | |
FQ214281.1 | - | 5 - 6 | 773 | 106 | ||
FQ214493.1 | - | 5 - 6 | 774 | 106 | ||
FQ219244.1 | - | 5 - 6 | 772 | 106 | ||
FQ219787.1 | - | 5 - 6 | 774 | 106 | ||
FQ219848.1 | - | 5 - 6 | 774 | 106 | ||
FQ220161.1 | - | 5 - 6 | 774 | 106 | ||
FQ220205.1 | - | 5 - 6 | 774 | 106 | ||
FQ220219.1 | - | 5 - 6 | 774 | 106 | ||
FQ227439.1 | - | 5 - 6 | 774 | 106 | ||
FQ229451.1 | - | 5 - 6 | 775 | 106 | ||
FQ229816.1 | - | 5 - 6 | 772 | 106 | ||
FQ230017.1 | - | 5 - 6 | 772 | 106 | ||
FQ230102.1 | - | 5 - 6 | 774 | 106 | ||
FQ230128.1 | - | 5 - 6 | 802 | 106 | ||
FQ230794.1 | - | 5 - 6 | 777 | 106 | ||
FQ231729.1 | - | 5 - 6 | 772 | 106 | ||
FQ232540.1 | - | 5 - 6 | 774 | 106 | ||
FQ233314.1 | - | 5 - 6 | 799 | 106 | ||
FQ234171.1 | - | 5 - 6 | 804 | 106 | ||
X06148.1 | - | 5 - 6 | 826 | 106 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
