Assay Details
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031620.1 | NP_113808.1 | 6 - 7 | 618 | 71 | |
XM_006233010.1 | XP_006233072.1 | 5 - 6 | 641 | 71 | ||
GenBank mRNA | BC086327.1 | - | 5 - 6 | 646 | 71 | |
X97772.1 | - | 5 - 6 | 577 | 71 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
