Assay Details
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 1 - 2 | 129 | 61 | |
GenBank mRNA | BC059125.1 | - | 1 - 2 | 57 | 61 | |
FQ213475.1 | - | 1 - 2 | 53 | 61 | ||
FQ225110.1 | - | 1 - 2 | 91 | 61 | ||
M34477.1 | - | 1 - 2 | 129 | 61 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
This assay is in the 5'UTR region of the BC059125.1, M34477.1, NM_031840.1 transcripts.
This assay is in the 5'UTR region of the BC059125.1, M34477.1, NM_031840.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
