Assay Details
Gene Symbol: Fdps | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031840.1 | NP_114028.1 | 3 - 4 | 441 | 92 | |
GenBank mRNA | BC059125.1 | - | 3 - 4 | 369 | 92 | |
FQ209697.1 | - | 3 - 4 | 334 | 92 | ||
FQ210400.1 | - | 3 - 4 | 333 | 92 | ||
FQ211073.1 | - | 4 - 5 | 550 | 92 | ||
FQ213475.1 | - | 3 - 4 | 365 | 92 | ||
FQ218913.1 | - | 3 - 4 | 335 | 92 | ||
FQ218933.1 | - | 3 - 4 | 335 | 92 | ||
FQ219092.1 | - | 3 - 4 | 335 | 92 | ||
FQ225110.1 | - | 3 - 4 | 403 | 92 | ||
M34477.1 | - | 3 - 4 | 441 | 92 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
