Assay Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053420.3 | NP_445872.1 | 4 - 5 | 492 | 60 | |
GenBank mRNA | AF243515.1 | - | 4 - 5 | 456 | 60 | |
AY095482.1 | - | 4 - 5 | 456 | 60 | ||
BC070958.1 | - | 4 - 5 | 450 | 60 | ||
BC081690.1 | - | 4 - 5 | 430 | 60 | ||
CO403082.1 | - | 4 - 5 | 528 | 60 | ||
FQ213448.1 | - | 3 - 4 | 253 | 60 | ||
FQ215108.1 | - | 4 - 5 | 462 | 60 | ||
FQ215127.1 | - | 4 - 5 | 462 | 60 | ||
FQ215203.1 | - | 4 - 5 | 483 | 60 | ||
FQ216745.1 | - | 4 - 5 | 483 | 60 | ||
FQ216768.1 | - | 1 - 2 | 69 | 60 | ||
FQ216807.1 | - | 4 - 5 | 483 | 60 | ||
FQ220853.1 | - | 4 - 5 | 461 | 60 | ||
FQ221849.1 | - | 4 - 5 | 483 | 60 | ||
KC660101.1 | - | 3 - 4 | 289 | 60 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
