Assay Details
Gene Symbol: Bnip3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053420.3 | NP_445872.1 | 5 - 6 | 640 | 108 | |
GenBank mRNA | AF243515.1 | - | 5 - 6 | 604 | 108 | |
AY095482.1 | - | 5 - 6 | 604 | 108 | ||
BC070958.1 | - | 5 - 6 | 598 | 7105707 | 108 | |
BC081690.1 | - | 5 - 6 | 578 | 7190121 | 108 | |
FQ215108.1 | - | 5 - 6 | 610 | 108 | ||
FQ215127.1 | - | 5 - 6 | 610 | 108 | ||
FQ215203.1 | - | 5 - 6 | 631 | 108 | ||
FQ216745.1 | - | 5 - 6 | 631 | 108 | ||
FQ216768.1 | - | 2 - 3 | 217 | 108 | ||
FQ220853.1 | - | 5 - 6 | 609 | 108 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
