Assay Details
Gene Symbol: Tdg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053729.1 | NP_446181.1 | 3 - 4 | 445 | 79 | |
GenBank mRNA | AY026945.1 | - | 3 - 4 | 445 | 79 | |
BC129088.1 | - | 3 - 4 | 462 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
