Assay Details
Gene Symbol: Vmp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_138839.2 | NP_620194.1 | 6 - 7 | 695 | 94 | |
XM_006247078.2 | XP_006247140.1 | 6 - 7 | 1173 | 94 | ||
XM_006247079.2 | XP_006247141.1 | 7 - 8 | 717 | 94 | ||
GenBank mRNA | AF411216.1 | - | 6 - 7 | 641 | 94 | |
BC061721.1 | - | 6 - 7 | 695 | 94 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
