Assay Details
Gene Symbol: Il13ra1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_145789.2 | NP_665732.2 | 10 - 11 | 1236 | 117 | |
GenBank mRNA | AY044251.1 | - | 10 - 11 | 1220 | 117 | |
BC093615.1 | - | 10 - 11 | 1236 | 117 |
Gene Symbol: LOC100360218 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_003751077.3 | XP_003751125.1 | 1 | 1262 | 117 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P389072 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P741723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619050 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 386305 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 558225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 751974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 25132 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492534 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 50345 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 816684 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 433518 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 850001 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 231238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 753233 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 332400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 276791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 475976 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 446229 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269619 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 60599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 517481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 728081 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 400134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83982 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 241434 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 15905 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 833862 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 878431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 58607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661396 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 536972 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 175784 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 52839 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 380142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 114252 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 126140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 286842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187481 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 561077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 586445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486690 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 128588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 97660 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8740 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 18657 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 168607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 41238 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 43798 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 20669 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P389072 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P741723 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P741723 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P389072 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P389072 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P741723 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P948250 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P389072 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P362915 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P741723 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 43541 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6229 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23132 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 14195 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 56788 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10936366 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936374 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754986 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936370 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936376 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754985 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936372 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936368 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936369 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936371 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936375 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754984 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936373 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936367 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10936365 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10754983 | - | - |
Rat Genome 230 2.0 Array | 1388711_PM_at | - | - |
Rat Genome 230 2.0 Array | 1370728_at | - | - |
Rat Genome 230 2.0 Array | 1388711_at | - | - |
Rat Genome 230 2.0 Array | 1370728_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1388711_at | - | - |
Affymetrix HT Rat Focus Array | 1370728_at | - | - |
Affymetrix HT Rat Focus Array | 1388711_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1370728_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00006459.rn.1 | TC0X00001043.rn.1 | RTA TC |
Rat Expression Set 230 | 1370728_PM_at | - | - |
Rat Expression Set 230 | 1388711_at | - | - |
Rat Expression Set 230 | 1388711_PM_at | - | - |
Rat Expression Set 230 | 1370728_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10936365 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10754983 | - | - |
Rat Genome U34 Set | rc_AI009602_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21067285 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_2040922 | - | - |
RatRef-12 Expression BeadChip | ILMN_66894 | - | - |
