Assay Details
Gene Symbol: Aurka | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_153296.2 | NP_695208.2 | 5 - 6 | 693 | 98 | |
XM_006235675.2 | XP_006235737.1 | 6 - 7 | 772 | 98 | ||
XM_006235676.2 | XP_006235738.1 | 6 - 7 | 789 | 98 | ||
GenBank mRNA | AF537333.1 | - | 5 - 6 | 681 | 98 | |
BC104676.1 | - | 5 - 6 | 693 | 98 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
