Assay Details
Gene Symbol: Acp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_021262.2 | NP_067085.1 | 3 - 4 | 272 | 118 | |
XM_006239965.2 | XP_006240027.1 | 3 - 4 | 301 | 118 | ||
GenBank mRNA | AF171072.1 | - | 3 - 4 | 235 | 118 | |
BC062229.1 | - | 3 - 4 | 272 | 118 | ||
FQ213063.1 | - | 3 - 4 | 252 | 118 | ||
FQ214806.1 | - | 2 - 3 | 140 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
