Assay Details
Gene Symbol: Srsf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001195505.1 | NP_001182434.1 | 631 | 102 | ||
NM_001195506.1 | NP_001182435.1 | 502 | 102 | |||
NM_019257.3 | NP_062130.2 | 506 | 102 | |||
XM_008764878.1 | XP_008763100.1 | 5 - 6 | 682 | 102 | ||
XM_008764879.1 | XP_008763101.1 | 5 - 6 | 553 | 102 | ||
GenBank mRNA | BC058479.1 | - | 500 | 102 | ||
L13635.1 | - | 602 | 102 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
