Assay Details
Gene Symbol: Syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006243515.1 | XP_006243577.1 | 2107 | 52 | ||
XM_006243519.2 | XP_006243581.1 | 10 - 11 | 1603 | 52 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
