Assay Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106006.1 | NP_001099476.1 | 1 - 2 | 274 | 122 | |
GenBank mRNA | FQ226880.1 | - | 1 - 2 | 266 | 122 | |
FQ233155.1 | - | 1 - 2 | 222 | 122 | ||
FQ234076.1 | - | 1 - 2 | 252 | 122 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
Function(s)
Process(es)
protein polyubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity
ubiquitin protein ligase activity
protein polyubiquitination
regulation of proteasomal ubiquitin-dependent protein catabolic process
proteasome-mediated ubiquitin-dependent protein catabolic process
intrinsic apoptotic signaling pathway in response to endoplasmic reticulum stress
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
ligase activity
ubiquitin-like protein transferase activity
ubiquitin protein ligase binding
ubiquitin-ubiquitin ligase activity
ubiquitin protein ligase activity
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P066003 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_42_P573232 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 731522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122817 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 171131 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 136723 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255373 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 189208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 504924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254995 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356565 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 941913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349912 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 330692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 944608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865848 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 478240 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 255756 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 54842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 417517 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143125 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 507508 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 671217 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 568902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900814 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 470578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959169 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 900579 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350911 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654707 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 99908 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865445 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 580304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 853509 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 912759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 654201 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 737184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 546127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 207179 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 405213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 823625 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 444942 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155714 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 612299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 71724 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 773642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302082 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 388067 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 636869 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 57154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 425477 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 116718 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 645863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 136436 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 112683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 49538 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 66537 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91461 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10977 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 35506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59600 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71053 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 166796 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 60910 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P573232 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066003 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P066003 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_42_P573232 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_42_P573232 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P066003 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_42_P842057 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9664 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 104485 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 23482 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 84369 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10776803 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776804 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776801 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776805 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776806 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776802 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10776800 | - | - |
Rat Genome 230 2.0 Array | 1385259_PM_at | - | - |
Rat Genome 230 2.0 Array | 1373063_at | - | - |
Rat Genome 230 2.0 Array | 1383515_at | - | - |
Rat Genome 230 2.0 Array | 1375965_at | - | - |
Rat Genome 230 2.0 Array | 1373063_PM_at | - | - |
Rat Genome 230 2.0 Array | 1385259_at | - | - |
Rat Genome 230 2.0 Array | 1383515_PM_at | - | - |
Rat Genome 230 2.0 Array | 1375965_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1385259_at | - | - |
Affymetrix HT Rat Focus Array | 1385259_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373063_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1373063_at | - | - |
Affymetrix HT Rat Focus Array | 1375965_at | - | - |
Affymetrix HT Rat Focus Array | 1375965_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1400013043.rn.1 | TC1400001773.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400013040.rn.1 | TC1400001773.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1400013041.rn.1 | TC1400001773.rn.1 | RTA TC |
Rat Expression Set 230 | 1375965_PM_at | - | - |
Rat Expression Set 230 | 1373063_PM_at | - | - |
Rat Expression Set 230 | 1375965_at | - | - |
Rat Expression Set 230 | 1373063_at | - | - |
Rat Expression Set 230 | 1383515_at | - | - |
Rat Expression Set 230 | 1385259_PM_at | - | - |
Rat Expression Set 230 | 1385259_at | - | - |
Rat Expression Set 230 | 1383515_PM_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10776800 | - | - |
Rat Genome U34 Set | rc_AA998447_at | - | - |
Rat Genome U34 Set | rc_AI059463_at | - | - |
Rat Genome U34 Set | rc_AI179857_at | - | - |
Rat Genome U34 Set | rc_AI229167_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21980381 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1370917 | - | - |
