Assay Details
Gene Symbol: Csnk1a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053615.1 | NP_446067.1 | 4 - 5 | 448 | 111 | |
GenBank mRNA | BC127486.1 | - | 674 | 111 | ||
U77582.1 | - | 4 - 5 | 448 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
