Assay Details
Gene Symbol: Dpf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108516.1 | NP_001101986.1 | 6 - 7 | 682 | 62 | |
GenBank mRNA | BC166788.1 | - | 6 - 7 | 629 | 62 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
