Assay Details
Gene Symbol: Prdx4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053512.2 | NP_445964.1 | 5 - 6 | 763 | 96 | |
XM_006256951.2 | XP_006257013.1 | 5 - 6 | 928 | 96 | ||
GenBank mRNA | AF106945.1 | - | 5 - 6 | 764 | 96 | |
BC059122.1 | - | 5 - 6 | 847 | 96 | ||
FQ219258.1 | - | 5 - 6 | 811 | 96 | ||
FQ220216.1 | - | 5 - 6 | 817 | 96 | ||
FQ220257.1 | - | 5 - 6 | 811 | 96 | ||
FQ220916.1 | - | 5 - 6 | 812 | 96 | ||
FQ228535.1 | - | 5 - 6 | 812 | 96 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
