Assay Details
Gene Symbol: Ammecr1l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001107399.1 | NP_001100869.1 | 7 - 8 | 1080 | 126 | |
XM_006254590.2 | XP_006254652.1 | 7 - 8 | 1894 | 126 | ||
XM_006254593.2 | XP_006254655.1 | 7 - 8 | 1331 | 126 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_64_P028335 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P329688 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 970294 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 524401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 485263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 521992 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496512 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 389191 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 119516 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 457370 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 847042 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625020 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 158571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 519866 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 932456 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 814772 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 959759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 94666 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 120383 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 13882 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P329688 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028335 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P329688 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_64_P028335 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_64_P028335 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P329688 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P260384 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P329688 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P148695 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10800659 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800658 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800663 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800666 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800660 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800665 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800662 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800664 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800661 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10800657 | - | - |
Rat Genome 230 2.0 Array | 1378894_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378338_at | - | - |
Rat Genome 230 2.0 Array | 1392488_at | - | - |
Rat Genome 230 2.0 Array | 1392488_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378338_PM_at | - | - |
Rat Genome 230 2.0 Array | 1378894_at | - | - |
Affymetrix HT Rat Focus Array | 1392488_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1392488_at | - | - |
Affymetrix HT Rat Focus Array | 1378338_at | - | - |
Affymetrix HT Rat Focus Array | 1378338_PM_at | - | - |
Affymetrix Rat Clariom S Assay | PSR1800001453.rn.1 | TC1800000216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800001451.rn.1 | TC1800000216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800001454.rn.1 | TC1800000216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800001452.rn.1 | TC1800000216.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR1800001450.rn.1 | TC1800000216.rn.1 | RTA TC |
Rat Expression Set 230 | 1392488_at | - | - |
Rat Expression Set 230 | 1392488_PM_at | - | - |
Rat Expression Set 230 | 1378338_PM_at | - | - |
Rat Expression Set 230 | 1378338_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10800657 | - | - |
Rat Genome U34 Set | rc_AA819359_at | - | - |
Rat Genome U34 Set | rc_AI168950_at | - | - |
Rat Genome U34 Set | rc_AI113288_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21987796 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1359345 | - | - |
