Assay Details
Gene Symbol: Ptbp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271057.1 | NP_001257986.1 | 10 - 11 | 1216 | 59 | |
NM_022516.5 | NP_071961.2 | 9 - 10 | 1138 | 59 | ||
GenBank mRNA | BC061858.1 | - | 10 - 11 | 1211 | 59 | |
X60789.1 | - | 9 - 10 | 1101 | 59 | ||
X60790.1 | - | 6 - 7 | 587 | 59 | ||
X74565.1 | - | 10 - 11 | 1197 | 59 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
