Assay Details
Gene Symbol: Prodh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001038588.1 | NP_001033677.1 | 1 - 2 | 184 | 57 | |
GenBank mRNA | AF222852.1 | - | 1 - 2 | 174 | 57 | |
BC158806.1 | - | 1 - 2 | 184 | 57 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
