Assay Details
Gene Symbol: Akt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017093.1 | NP_058789.1 | 6 - 7 | 850 | 81 | |
XM_008759108.1 | XP_008757330.1 | 7 - 8 | 3249 | 81 | ||
XM_008759109.1 | XP_008757331.1 | 6 - 7 | 837 | 81 | ||
XM_008759110.1 | XP_008757332.1 | 6 - 7 | 3114 | 81 | ||
XM_008759111.1 | XP_008757333.1 | 6 - 7 | 960 | 81 | ||
GenBank mRNA | D30041.1 | - | 6 - 7 | 850 | 81 | |
DQ198085.1 | - | 5 - 6 | 569 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
