Assay Details
Gene Symbol: Ap1m2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108996.2 | NP_001102466.1 | 3 - 4 | 305 | 74 | |
GenBank mRNA | BC167082.1 | - | 3 - 4 | 305 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
