Assay Details
Gene Symbol: Shmt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008322.1 | NP_001008323.1 | 2 - 3 | 283 | 111 | |
XM_006241453.2 | XP_006241515.1 | 2 - 3 | 309 | 111 | ||
GenBank mRNA | BC085331.1 | - | 2 - 3 | 283 | 111 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
