Assay Details
Gene Symbol: Cbl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_576396.6 | XP_576396.3 | 9 - 10 | 1522 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() |
Gene Ontology Categories:
