Assay Details
Gene Symbol: U2af1l4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001008775.1 | NP_001008775.1 | 5 - 6 | 285 | 68 | |
XM_006228776.2 | XP_006228838.1 | 4 - 5 | 302 | 68 | ||
XM_006228777.1 | XP_006228839.1 | 3 - 4 | 186 | 68 | ||
XM_006228778.2 | XP_006228840.1 | 4 - 5 | 302 | 68 | ||
GenBank mRNA | AY325237.1 | - | 347 | 68 | ||
AY325238.1 | - | 5 - 6 | 285 | 68 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
