Assay Details
Gene Symbol: Aldh1a2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_053896.2 | NP_446348.2 | 1 - 2 | 119 | 65 | |
GenBank mRNA | BC098910.1 | - | 1 - 2 | 111 | 65 | |
BE111477.1 | - | 1 - 2 | 119 | 65 | ||
U60063.1 | - | 1 - 2 | 115 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
