Assay Details
Gene Symbol: Dlgap3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001276304.1 | NP_001263233.1 | 3 - 4 | 1445 | 91 | |
XM_008763989.1 | XP_008762211.1 | 4 - 5 | 2747 | 91 | ||
GenBank mRNA | AY530298.1 | - | 2 - 3 | 1308 | 91 | |
AY530299.1 | - | 2 - 3 | 1308 | 91 | ||
U67139.1 | - | 3 - 4 | 1412 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
