Assay Details
Gene Symbol: Fam3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001109324.1 | NP_001102794.1 | 7 - 8 | 773 | 65 | |
XM_006229603.2 | XP_006229665.1 | 6 - 7 | 930 | 65 | ||
XM_006229604.2 | XP_006229666.1 | 6 - 7 | 3342 | 65 | ||
XM_006229605.2 | XP_006229667.1 | 7 - 8 | 2655 | 65 | ||
GenBank mRNA | BC169081.1 | - | 6 - 7 | 837 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
