Assay Details
Gene Symbol: Nprl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001034936.1 | NP_001030108.1 | 3 - 4 | 418 | 64 | |
XM_006246008.2 | XP_006246070.1 | 4 - 5 | 540 | 64 | ||
XM_006246009.2 | XP_006246071.1 | 4 - 5 | 540 | 64 | ||
XM_006246010.2 | XP_006246072.1 | 3 - 4 | 500 | 64 | ||
XM_006246011.2 | XP_006246073.1 | 4 - 5 | 352 | 64 | ||
XM_006246013.2 | XP_006246075.1 | 2 - 3 | 1982 | 64 | ||
XM_006246014.2 | XP_006246076.1 | 2 - 3 | 1883 | 64 | ||
XM_006246015.2 | XP_006246077.1 | 2 - 3 | 2184 | 64 | ||
XM_006246016.2 | XP_006246078.1 | 4 - 5 | 552 | 64 | ||
GenBank mRNA | BC099803.1 | - | 3 - 4 | 418 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
