Assay Details
Gene Symbol: Cdc42 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_171994.4 | NP_741991.3 | 4 - 5 | 362 | 74 | |
XM_008764286.1 | XP_008762508.1 | 4 - 5 | 867 | 74 | ||
XM_008764287.1 | XP_008762509.1 | 4 - 5 | 442 | 74 | ||
GenBank mRNA | AF205635.1 | - | 3 - 4 | 289 | 74 | |
AF491841.1 | - | 3 - 4 | 355 | 74 | ||
BC060535.1 | - | 4 - 5 | 362 | 74 | ||
FQ213007.1 | - | 4 - 5 | 361 | 74 | ||
FQ225045.1 | - | 4 - 5 | 361 | 74 | ||
FQ225541.1 | - | 4 - 5 | 474 | 74 | ||
FQ225902.1 | - | 4 - 5 | 361 | 74 | ||
FQ225973.1 | - | 4 - 5 | 474 | 74 | ||
FQ226943.1 | - | 4 - 5 | 357 | 74 | ||
FQ227167.1 | - | 4 - 5 | 361 | 74 | ||
FQ227499.1 | - | 4 - 5 | 361 | 74 | ||
FQ227894.1 | - | 4 - 5 | 361 | 74 | ||
FQ228149.1 | - | 4 - 5 | 474 | 74 | ||
FQ230725.1 | - | 4 - 5 | 370 | 74 | ||
FQ231089.1 | - | 4 - 5 | 361 | 74 | ||
FQ231482.1 | - | 4 - 5 | 370 | 74 | ||
FQ231846.1 | - | 4 - 5 | 474 | 74 | ||
FQ232179.1 | - | 4 - 5 | 361 | 74 | ||
FQ232295.1 | - | 4 - 5 | 370 | 74 | ||
FQ232459.1 | - | 4 - 5 | 361 | 74 | ||
FQ233586.1 | - | 4 - 5 | 372 | 74 | ||
FQ234639.1 | - | 4 - 5 | 370 | 74 | ||
FQ234830.1 | - | 4 - 5 | 370 | 74 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
