Assay Details
Gene Symbol: Psma5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC060575.1 | - | 1 - 2 | 135 | 96 | |
FQ218317.1 | - | 1 - 2 | 131 | 96 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC060575.1 transcripts.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
