Assay Details
Gene Symbol: Elavl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001108848.1 | NP_001102318.1 | 1 - 2 | 136 | 86 | |
GenBank mRNA | BC168972.1 | - | 1 - 2 | 142 | 86 |

More Information
Updated Mapping Information

Additional Information:
This assay is in the 5'UTR region of the BC168972.1, NM_001108848.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
