Assay Details
Gene Symbol: Ube2k | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001106006.1 | NP_001099476.1 | 3 - 4 | 428 | 130 | |
XM_006250934.2 | XP_006250996.1 | 3 - 4 | 625 | 130 | ||
GenBank mRNA | FQ226880.1 | - | 3 - 4 | 420 | 130 | |
FQ233155.1 | - | 3 - 4 | 376 | 130 | ||
FQ234076.1 | - | 3 - 4 | 406 | 130 |

More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
