Assay Details
Gene Symbol: Actr1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039028.2 | NP_001034117.2 | 1 - 2 | 156 | 71 | |
GenBank mRNA | BC166803.1 | - | 1 - 2 | 123 | 71 | |
EV779654.1 | - | 1 - 2 | 184 | 71 | ||
FQ228979.1 | - | 1 - 2 | 124 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
