Assay Details
Gene Symbol: Cnih2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025132.1 | NP_001020303.1 | 2 - 3 | 401 | 79 | |
XM_008760153.1 | XP_008758375.1 | 1 - 2 | 979 | 79 | ||
XM_008760154.1 | XP_008758376.1 | 2 - 3 | 1009 | 79 | ||
GenBank mRNA | BC091325.1 | - | 2 - 3 | 401 | 79 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
