Assay Details
Gene Symbol: Cpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC085762.1 | - | 8 - 9 | 1418 | 85 | |
J04625.1 | - | 8 - 9 | 1374 | 85 | ||
M31602.1 | - | 8 - 9 | 1390 | 85 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
MicroArray Probes:
