Assay Details
Gene Symbol: Gps2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017477.1 | NP_001017477.1 | 5 - 6 | 764 | 65 | |
XM_006246783.2 | XP_006246845.1 | 6 - 7 | 714 | 65 | ||
XM_006246785.1 | XP_006246847.1 | 6 - 7 | 714 | 65 | ||
XM_008767855.1 | XP_008766077.1 | 6 - 7 | 714 | 65 | ||
XM_008767856.1 | XP_008766078.1 | 6 - 7 | 714 | 65 | ||
GenBank mRNA | BC083699.1 | - | 5 - 6 | 764 | 65 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
