Assay Details
Gene Symbol: Acbd4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012013.1 | NP_001012013.1 | 2 - 3 | 367 | 73 | |
XM_006247508.2 | XP_006247570.1 | 3 - 4 | 355 | 73 | ||
XM_006247509.2 | XP_006247571.1 | 3 - 4 | 357 | 73 | ||
XM_006247510.2 | XP_006247572.1 | 3 - 4 | 356 | 73 | ||
XM_006247511.2 | XP_006247573.1 | 3 - 4 | 355 | 73 | ||
XM_006247512.2 | XP_006247574.1 | 3 - 4 | 356 | 73 | ||
XM_008768284.1 | XP_008766506.1 | 4 - 5 | 458 | 73 | ||
XM_008768285.1 | XP_008766507.1 | 3 - 4 | 346 | 73 | ||
XM_008768286.1 | XP_008766508.1 | 3 - 4 | 357 | 73 | ||
XM_008768287.1 | XP_008766509.1 | 3 - 4 | 356 | 73 | ||
GenBank mRNA | BC076387.1 | - | 2 - 3 | 367 | 73 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
