Assay Details
Gene Symbol: Bhlhe41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002729454.5 | XP_002729500.2 | 4 - 5 | 687 | 66 | |
GenBank mRNA | AF009329.1 | - | 4 - 5 | 585 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
