Assay Details
Gene Symbol: Mob4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_133528.1 | NP_598212.1 | 6 - 7 | 442 | 89 | |
GenBank mRNA | AJ132008.1 | - | 6 - 7 | 442 | 89 | |
BC085708.1 | - | 6 - 7 | 463 | 89 | ||
FQ229796.1 | - | 6 - 7 | 458 | 89 | ||
FQ233766.1 | - | 6 - 7 | 465 | 89 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
