Assay Details
Gene Symbol: Maged2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001271109.1 | NP_001258038.1 | 12 - 13 | 1927 | 64 | |
NM_001271110.1 | NP_001258039.1 | 12 - 13 | 1955 | 64 | ||
XM_006237924.2 | XP_006237986.1 | 12 - 13 | 2017 | 64 | ||
GenBank mRNA | AJ293617.1 | - | 11 - 12 | 1868 | 64 | |
AY724507.1 | - | 12 - 13 | 1940 | 64 | ||
BC086998.1 | - | 10 - 11 | 2884 | 64 | ||
FQ229095.1 | - | 12 - 13 | 1939 | 64 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
