Assay Details
Gene Symbol: Nabp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001014216.1 | NP_001014238.1 | 3 - 4 | 496 | 93 | |
GenBank mRNA | BC089852.1 | - | 3 - 4 | 496 | 93 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
