Assay Details
Gene Symbol: Phgdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_031620.1 | NP_113808.1 | 10 - 11 | 1192 | 71 | |
XM_006233010.1 | XP_006233072.1 | 9 - 10 | 1215 | 71 | ||
GenBank mRNA | BC086327.1 | - | 9 - 10 | 1220 | 71 | |
X97772.1 | - | 9 - 10 | 1151 | 71 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
