Assay Details
Gene Symbol: Asic4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022234.2 | NP_071570.2 | 3 - 4 | 1166 | 80 | |
XM_006245275.2 | XP_006245337.1 | 4 - 5 | 1295 | 80 | ||
GenBank mRNA | AJ242554.1 | - | 3 - 4 | 1147 | 80 | |
AJ271642.1 | - | 3 - 4 | 80 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
