Assay Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017374.2 | NP_001017374.2 | 20 - 21 | 2836 | 149 | |
GenBank mRNA | AB256044.1 | - | 6 - 7 | 1063 | 149 | |
AM258959.1 | - | 6 - 7 | 1063 | 149 | ||
BC091330.1 | - | 6 - 7 | 1147 | 149 | ||
BC099117.1 | - | 20 - 21 | 2723 | 149 | ||
FQ232649.1 | - | 3 - 4 | 468 | 149 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 149 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
