Assay Details
Gene Symbol: Eif4g2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001017374.2 | NP_001017374.2 | 8 - 9 | 1000 | 107 | |
GenBank mRNA | BC099117.1 | - | 8 - 9 | 887 | 107 |
Gene Symbol: LOC501116 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008765067.1 | XP_008763289.1 | 2 | 822 | 107 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
