Assay Details
Gene Symbol: Ralyl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_006232123.2 | XP_006232185.1 | 6 - 7 | 844 | 71 | |
XM_008760852.1 | XP_008759074.1 | 6 - 7 | 844 | 71 | ||
XM_008760853.1 | XP_008759075.1 | 6 - 7 | 1341 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P236448 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 161127 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 28964 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47005 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 295089 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459211 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642320 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 803974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 649998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 552607 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 412961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 874502 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 841802 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 972672 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 254790 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384643 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 384726 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 47956 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522180 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 39703 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 917326 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 350913 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 155189 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 953184 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 23088 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 239967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 886553 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 37773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 106924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93301 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 250142 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 744198 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667693 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530284 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3154 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 935506 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 83650 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 115228 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 633148 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 287463 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 294371 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 319483 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961971 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419896 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 55314 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 59070 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734234 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 788185 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390105 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 215374 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181750 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 266832 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592809 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 164297 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 24654 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718528 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 203168 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 327787 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202442 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 419077 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459570 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718186 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 786152 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 622114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 542921 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 650855 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 103677 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 727661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 149641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 600962 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 379158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 668025 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 794917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107924 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 747623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 768106 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 459414 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 916361 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 497196 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 147317 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 486423 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 196573 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 865653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 963266 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 349271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 304720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 399759 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 706328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 97931 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 755085 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 468522 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 663084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 258330 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 966835 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174479 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 110062 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 395177 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 806328 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 734953 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 187083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 141218 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 173773 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77588 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 472743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 503225 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 31762 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 596941 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 588651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 562497 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 828661 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 891955 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 117312 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 184641 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 679401 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 98632 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302149 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 202545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 973864 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 89032 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 211475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 743729 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 309230 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 525595 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 451417 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 845492 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 699188 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 435904 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 592084 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 801751 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945321 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 382140 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 697980 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 387472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 619133 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 3420 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 482824 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 77475 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 721097 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 345304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262567 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 951272 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720807 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 946384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 957268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 156602 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 246523 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905806 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 807531 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 856224 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143583 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 890521 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247771 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 926575 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 680278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 771104 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 492967 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 458439 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 538413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14535 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 236589 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 677274 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290651 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756011 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 739873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 165174 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 38422 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 99675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 87960 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 59079 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 52858 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164271 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 167928 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89248 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 71586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 8681 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 148987 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 89692 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151431 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11932 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63208 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63440 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 11259 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163056 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 141757 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 116433 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 105810 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 64378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91413 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 54721 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 175114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 33753 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163667 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 158653 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 10470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 103792 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 134719 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236448 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P236448 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P236448 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P236448 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10822309 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822306 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822313 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822311 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822312 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10822305 | - | - |
Rat Genome 230 2.0 Array | 1380852_PM_at | - | - |
Rat Genome 230 2.0 Array | 1397513_at | - | - |
Rat Genome 230 2.0 Array | 1380852_at | - | - |
Rat Genome 230 2.0 Array | 1397513_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380852_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1380852_at | - | - |
Rat Expression Set 230 | 1397513_PM_at | - | - |
Rat Expression Set 230 | 1397513_at | - | - |
Rat Expression Set 230 | 1380852_PM_at | - | - |
Rat Expression Set 230 | 1380852_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10822305 | - | - |
Rat Genome U34 Set | rc_AI112346_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1353565 | - | - |
