Assay Details
Gene Symbol: Tomm40l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083338.1 | NP_001076807.1 | 3 - 4 | 277 | 90 | |
XM_006250247.2 | XP_006250309.1 | 4 - 5 | 756 | 90 | ||
GenBank mRNA | AB274732.1 | - | 3 - 4 | 277 | 90 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() |
Gene Ontology Categories:
MicroArray Probes:
