Assay Details
Gene Symbol: Gabra3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_017069.3 | NP_058765.3 | 4 - 5 | 564 | 81 | |
XM_006229511.2 | XP_006229573.1 | 4 - 5 | 676 | 81 | ||
GenBank mRNA | CB556618.1 | - | 3 - 4 | 392 | 81 | |
X51991.1 | - | 4 - 5 | 571 | 81 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Whole Rat Genome Microarray 4x44K v2 | A_44_P535601 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 298876 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 533635 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 688920 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864683 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 945451 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 427215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 437675 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 107083 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 84548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956438 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 284412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595158 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 2775 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 545919 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 782823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 93733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 725548 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 420503 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 192046 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 134499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 931733 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 956419 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 516182 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 657219 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 929048 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 530586 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 496313 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 653329 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343165 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 32210 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 385123 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 46711 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 168486 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 854491 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 343339 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377120 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877143 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 205213 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 825263 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 123823 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 247437 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968074 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 557362 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964430 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 181024 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 267412 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 445863 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 640841 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 741922 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 366304 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 290614 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 17279 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 170917 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 174160 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 661634 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 512830 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 556260 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 244865 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 798513 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 625867 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 79671 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 593623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 595585 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 94254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 505299 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 53278 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 506378 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 356066 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 269060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 762026 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 756400 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 44087 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 875783 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 897360 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 483755 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 613612 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 522974 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 150076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 450203 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 864333 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 766392 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 750384 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 563743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 14998 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 6915 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 201465 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 143720 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 381245 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 617828 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 377550 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 964662 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 553545 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 476732 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 792576 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 390296 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 877581 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 302470 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 785311 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 56842 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 621948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 683788 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 955791 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 905049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 369608 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 718961 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 122268 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 333484 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 968325 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 120049 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 481071 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 262624 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 685558 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 62599 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 710134 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 839578 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 229571 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 767623 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 961743 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 212731 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 888059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 590254 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 316882 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 862151 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898114 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 642958 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 863027 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 681244 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 720499 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 667215 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 764800 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 852642 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 898402 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396873 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 396307 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 429948 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 12592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 1x1M | 7926 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 6929 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 91584 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 81009 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 63247 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 27669 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 164592 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 180101 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 90879 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 15170 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 39059 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 61231 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 79076 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114822 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147387 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 28007 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 74287 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 151472 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 85275 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 106986 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 31178 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 51327 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 147453 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 140902 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 133826 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 34060 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 30352 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 50153 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 114734 | - | - |
Agilent SurePrint G3 Rat CGH Microarray 4x180K | 163814 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P535601 | - | - |
Agilent Whole Rat Genome Microarray 4x44K v3 | A_44_P535601 | - | - |
Agilent SurePrint G3 Rat GE 8x60K Microarray | A_44_P535601 | - | - |
SurePrint G3 Rat Gene Expression Microarray G4131A | A_44_P535601 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 57972 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 9307 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 44205 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 91832 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 50402 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90659 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 74504 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 3524 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 68095 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 90497 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 6453 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7810 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 7611 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 38171 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 96770 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 35505 | - | - |
Agilent Rat Genome CGH Microarray 105A (G4436A) | 70032 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Rat Gene 1.0 ST Array | 10940095 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940092 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940093 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940094 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940097 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940091 | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940096 | - | - |
Affymetrix Rat Gene 1.1 ST Array | 10940090 | - | - |
Rat Genome 230 2.0 Array | 1369844_PM_at | - | - |
Rat Genome 230 2.0 Array | 1369844_at | - | - |
Affymetrix HT Rat Focus Array | 1369844_PM_at | - | - |
Affymetrix HT Rat Focus Array | 1369844_at | - | - |
Affymetrix Rat Clariom S Assay | PSR0X00015988.rn.1 | TC0X00002592.rn.1 | RTA TC |
Affymetrix Rat Clariom S Assay | PSR0X00015989.rn.1 | TC0X00002592.rn.1 | RTA TC |
Rat Expression Set 230 | 1369844_PM_at | - | - |
Rat Expression Set 230 | 1369844_at | - | - |
Affymetrix Rat Gene 1.0 ST Array | 10940090 | - | - |
Rat Genome U34 Set | X51991_at | - | - |
Rat Genome U34 Set | L08492cds_s_at | - | - |
Rat Genome U34 Set | L08492cds_s_at | - | - |
Rat Genome U34 Set | X51991_at | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Rat Genome Survey Microarray | 21550045 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Illumina ratRef-12 v1.0 expression beadchip | ILMN_1358002 | - | - |
RatRef-12 Expression BeadChip | ILMN_67998 | - | - |
